Hot plates, SG, SGR and SGK series
Lieferant: GESTIGKEIT HARRY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
SGK1 SGK6 SG3 SG5 SGR3 SGK4 SGR4 SG2 SG6 SGR2
GESTSGK1EA 306 EUR InStock
GESTSGK1 GESTSGK6 GESTSG3 GESTSG5 GESTSGR3 GESTSGK4 GESTSGR4 GESTSG2 GESTSG6 GESTSGR2
Hot plates, SG, SGR and SGK series
Heizgeräte Heizplatten
These hotplates made from smoothly finished cast-iron have a concave central expansion recesses. They are used for heating units which have a screw-in fitting for a supporting retort rod and have been designed for continuous operation .
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge