Electrical sandbaths, LHG
Lieferant: GESTIGKEIT HARRY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
ST72 ST82 ST92-230 ST73 SD8 SD7 ST71 ST92-400 ST83 SD9-230 ST81 ST91 ST93-400 ST93-230
453-2211EA 1240 EUR InStock
453-2211 453-2212 453-2213 453-2214 453-0040 453-0039 GESTST71 GESTST92-400 GESTST83 453-0089 GESTST81 GESTST91 GESTST93-400 GESTST93-230
Electrical sandbaths, LHG
Heizgeräte Sandbäder
Aluminium alloy plate, stainless steel housing with variable temperature control.
  • Robust construction for continuous performance
  • Even temperature is guaranteed over the entire heating surface
Sealed stainless steel frame with a usable height of 50 mm. The sand lays directly in contact with the hotplate. Four adjustable feet ensure extra stability.
Achtung! Bitte beachten: With thermostatic regulator.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge