Positive pressure manifold, ASPEC®
Lieferant: GILSON LCMS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
37012000
GILI37012000EA 4100 EUR InStock
GILI37012000
Positive pressure manifold, ASPEC®
Verteiler
The ASPEC® Positive pressure manifold uses positive pressure to improve reproducibility and recovery for solid phase extraction (SPE) applications.
  • Compact and easy to set up for quick operation
  • Positive pressure for reproducible extractions
  • Parallel processing for maximum efficiency
  • Same unit for 1, 3, 6 ml and 96 well formats
  • Compatible with viscous samples
  • Smart design to reduce waste management
With its precise flow rate control, the manifold improves consistency among samples and ensures uniform flow for both low and high viscosity solutions. Combined with ASPEC SPE Cartridges, the manifold offers an initial step to SPE automation for clinical, forensic, and food or beverage testing labs. With the ability to accommodate 96-well plate formats and viscous samples, it is also an ideal choice for sample clean up prior to LCMS for bioanalyses.
With its interchangeable manifolds, the ASPEC Manifold is compatible with both cartridges and 96-well SPE plates. Waste drain allows use of unlimited solvent volumes. Low gas pressure inlet (15 psi). Dual flow regulators for precise flow regulation for each SPE step. Combine with Gilson repetitive pipettes for accurate volume dispensing.
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge