Pipette controller, MACROMAN™
Lieferant: GILSON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F110120
GILSF110120EA 433 EUR InStock
GILSF110120
Pipette controller, MACROMAN™
Pipetten Pipettierhelfer
Pipette controller for all plastic and glass pipettes from 1 to 100 ml.
  • Ergonomic handle featuring wide triggers for maximum comfort and stress-free pipetting
  • Lithium-Ion battery enables a longer, vibration-free use with reduced charging time
  • Increase precision by choosing your flow rate mode from slow drops to rapid dispensing
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge