Syringe tips, REPET-TIPS, for electronic REPETMAN® pipettes
Lieferant: GILSON
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
F164558
GILSF164558EA 211 EUR InStock
GILSF164558
Syringe tips, REPET-TIPS, for electronic REPETMAN® pipettes
Pipettenspitzen
These tips are recommended for use with the electronic repetitive pipettor REPETMAN® when handling viscous, dense, volatile or hot liquids.
  • Automatic tip size recognition upon insertion
  • Free of dyes, no risk of contamination from the components
  • Sterilised tips are individually wrapped and certified free of endotoxins
Order Now
PRODUKT
TECHNISCHE DATEN
ZUBEHÖR
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge