Designed for performance and comfort, PIPETMAN® L comes in a comprehensive range of models, adapted to your needs in the lab. PIPETMAN L air-displacement pipettes cover a full volume range. Comfortable, precise, accurate, and reliable, use PIPETMAN L pipettes confidently in your applications.
- Convenient use
- Secure pipetting
- GLP compliance
Lightweight and balanced, PIPETMAN L combines an ergonomic design with minimized pipetting forces, giving you the ability to pipette longer without fatigue. Right-handed or left-handed? No problem. Adjust the location of the tip ejector to the most comfortable position for you.
PIPETMAN L single channel models are equipped with a stainless-steel tip ejector. Should your application require to pipette corrosive liquids, plastic tip ejectors are available as accessories. P5000L and P10mLL models are equipped with a plastic tip ejector to avoid direct contact with the tip and provide better compatibility with chemicals.
Reduce pipetting errors and avoid accidental volume changes during pipetting cycles with a true volume-locking mechanism for improved accuracy and precision. PIPETMAN L models are all fully autoclavable without disassembly for maximum convenience and protection from cross-contamination.
PIPETMAN L models feature a DataMatrix 2D code for easy traceability. Use a 2D code scanner to transfer the code automatically into any traceability or asset management system. Also, the pipette serial number is engraved on the body of the pipette for unique identification.
Lieferumfang: Das Mikrovolumen-Starter-Kit enthält die drei Pipetten P2L (0,2 - 2 µl), P10L (1 - 10 µl) und P100L (10 - 100 µl), zwei Racks mit DL10 Und D200 DIAMOND-Spitzen (jeweils 96 Spitzen), drei SINGLE™ Pipettenhalter, Inspektionsplakat, Gilson Anleitung zum Pipettieren und drei Kunststoff-Abwerfer.
Das Pipetman® L Starter-Kit enthält die drei Pipetten P20L (2 - 20 µl), P200L (20 - 200 µl), P1000L (100 - 1000 µl), zwei Racks mit D200 und D1000 DIAMOND-Spitzen (jeweils 96 Spitzen), drei SINGLE™-Pipettenhalter, Inspektionsplakat, Gilson Anleitung zum Pipettieren und drei Kunststoff-Abwerfer.
Der Pipetman® L 4-Pipetten-Kit enthält die vier Pipetten P2L (0,2 - 2 µl), (P20L (2 - 20 µl), P200L (20 - 200 µl), P1000L (100 - 1000 µl) mit Edelstahlabwurf, drei Racks mit DL10, D200 und D1000 DIAMOND-Spitzen (jeweils 96 Spitzen), vier SINGLE™-Pipettenhalter, Inspektionsplakat und Gilson Anleitung zum Pipettieren.