Nuboid wax
Lieferant: GLASS PRECISION ENGINEERING
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
70204 70206
GLPR70204EA 79.9 EUR InStock
GLPR70204 GLPR70206
Nuboid wax
Schmiermittel
Nuboid Waxes are high temperature lubricants for glass manufacturers and glassblowers.
The wax contains a specially selected grade of colloidal graphite and an agent which imports wetting and penetrating powers. They are all of high melting point and therefore do not soften until actually applied, they are also clean to use and reduce wastage to a minimum.The wax is designed for glassworking mould treatment and mould storage, automatic glass production, high temperature lubrication of rollers, shears and dies and for standard glassblowing.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge