Racks for upright freezers, slide boxes
Lieferant: GLW
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
K303 K304 K305 K306 K307 K308 K309
GLWAK303EA 277 EUR InStock
GLWAK303 GLWAK304 GLWAK305 GLWAK306 GLWAK307 GLWAK308 GLWAK309
Racks for upright freezers, slide boxes
Gestelle Kryogestelle
These freezer racks are made of stainless steel that are used for all common upright freezers available in different heights.
  • Drawers can hold upto 3 to 6 slide boxes
  • Each drawer is easily removable and has an ergonomic handle
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge