Overalls with collar, PP
Lieferant: GM EQUIPEMENT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
OJC036 OJC037 OJC035
GMEQOJC036EA 147 EUR InStock
GMEQOJC036 GMEQOJC037 GMEQOJC035
Overalls with collar, PP
Einweg-Kleidung
These disposable overalls are made of non woven PP.
  • Elasticated waist, cuffs and ankles
  • With collar and zip closure
  • Without pockets
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge