Frocks, PP
Lieferant: GM EQUIPEMENT
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
S732094
GMEQS732094EA 177 EUR InStock
GMEQS732094
Frocks, PP
Einweg-Kleidung
Disposable frock made of non-woven PP.
  • Back fastening: Bonds in back, neck and waist
  • Elasticated cuffs
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge