GOJO® Hand and Surface Scrubbing Wipes
Lieferant: GOJO Industries - Europe Ltd
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
9680-06-EEU
GOJO9680-06-EEUEA 132 EUR InStock
GOJO9680-06-EEU
GOJO® Hand and Surface Scrubbing Wipes
Wischtücher Wisch- und Handtücher, Einweg Desinfektionstücher
Hardworking hands need a hardworking wipe. Tough on everything from oil and grease to petrol and diesel, GOJO® hand and surface scrubbing wipes can be used anywhere heavy-duty hand and surface cleaning is required.
  • Dual-textured wet wipes with an abrasive side to remove heavy soils and a smooth side that leaves hands soft
  • Extra-large towels for cleaning hands, arms, tools, and surfaces
  • Cleaning and degreasing formula that has good skin compatibility
For use anywhere heavy-duty cleaning is required, with no water needed. Use in a variety of markets, including construction, manufacturing, and automobile industries.
Zertifizierungen: Complies with cosmetics regulation (regulation (EC) No 1223/2009) for use on hands and detergent regulation (regulation (EC) No 648/2004) for use on surfaces.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge