Jerricans, square or rectangular, HDPE, Corning® Gosselin™
Lieferant: Corning
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
HJC5-01
GOSSHJC5-01EA 280 EUR InStock
GOSSHJC5-01
Jerricans, square or rectangular, HDPE, Corning® Gosselin™
Flaschen Kanister
HDPE, translucent.
  • Good chemical resistance
  • Temperature resistant from −40 to +110 °C
Universal jerricans for industry, trade and laboratories. Ideal for packaging, storage and transportation.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge