Cell culture tube, CELLSTAR®, CELLreactor™
Lieferant: GREINER BIO ONE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
227245_500
GREI227245_500EA 780 EUR InStock
GREI227245_500
Cell culture tube, CELLSTAR®, CELLreactor™
Röhrchen Kulturröhrchen
PP, graduated, sterile, with filter screw cap
  • Facilitates a high number of parallel experiments
  • Flexible working volume
  • Maximal sterility and excellent gas exchange
  • Conical tube design and in-tube harvest
The CELLSTAR® CELLreactor™ can be used as small bioreactor for the cultiviation of suspension cells. In addition to cell culture applications the CELLSTAR® CELLreactor™ can be applied for the expansion of aerobic bacteria, yeast or other microorganisms in shaken cultures* as well as storage of components and liquids requiring gas exchange.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge