Sapphire Strips of 8 PCR Tube with Snap Strip Dome Cap
Lieferant: GREINER BIO ONE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
673283_1200
GREI673283_1200EA 764 EUR InStock
GREI673283_1200
Sapphire Strips of 8 PCR Tube with Snap Strip Dome Cap
Röhrchen PCR-Gefäße
Sapphire PCR 8-tube strip, breakable strips, 0,2 ml, with individual snap strip dome cap.
  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Temperature range: −20 to +105 °C
  • Non autoclavable
  • Shelf life: 5 years
  • Sterility: Non-sterile
Comprehensive thermal cycler compatibility.
Achtung! Bitte beachten: For single use only.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge