1536-well microplates, cyclo olefin
Lieferant: GREINER BIO ONE
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
792870-906
GREI792870-906EA 2070 EUR InStock
GREI792870-906
1536-well microplates, cyclo olefin
Mikrotiterplatten HTS-Mikrotiterplatten
Clear, 1536-well cycloolefin storage plates (with optimised geometry for low evaporation) for acoustic liquid handling.
  • Free from DNase, RNase, and human DNA
  • Groove around the plate edge matches cycloolefin plate lid (not included) - designed to reduce evaporation and minimise edge effects
Acoustic liquid dispensing offers the advantage of pipetting small volumes very precisely and with minimal disturbance without the risk of cross-contamination. The process involves the transferral of DMSO-containing solutions in the nanoliter to picoliter range from a starting plate into a target plate by means of ultrasound.
Zertifizierungen: Manufactured under DIN ISO 9001 guidelines.
Bestellinformation: Plates are supplied without lids. Accessory lid is sold separately.
Order Now
PRODUKT
TECHNISCHE DATEN
WEITERE INFORMATIONEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge