Digital vacuum - / barometer, GMH3161-12
Lieferant: Greisinger
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
600407
GRSG600407EA 0 EUR InStock
GRSG600407
Digital vacuum - / barometer, GMH3161-12
Barometer
This handheld meter with integrated absolute pressure sensor is suitable for measurements in air and non corrosive and non ionizing gases and liquids.
  • Impact resistant ABS, membrane keyboard, transparent panel, front side with IP65 rating
  • Min./Max. memory, HOLD function, automatic power-off
  • Sea level correction
  • Piezo-resistive absolute pressure sensor integrated in device
  • Serial interface (3,5 mm jack) can be connected to RS232 or USB interface of a PC via electrically isolated interface adapter (needs to be ordered separately)
Lieferumfang: Supplied with batteries and manual.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge