Coronavirus and SARS-CoV-2 RT real-time PCR kits, GSD NovaPrime®
Lieferant: GOLD STANDARD DIAGNOSTICS
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
PCOV6033 PCOV6034 PCOV6036 PCOV6037
GSDCPCOV6033EA 641 EUR InStock
GSDCPCOV6033 GSDCPCOV6034 GSDCPCOV6036 GSDCPCOV6037
Coronavirus and SARS-CoV-2 RT real-time PCR kits, GSD NovaPrime®
Klinisches Diagnosezubehör Klinische Reagenzien und Kits
This RT-PCR assay is a highly sensitive Multiplex Real-Time PCR with CE-IVD marking for the direct qualitative pathogen detection of SARS-CoV-2 genomic RNA in respiratory samples.
  • Approximately 1 hour turnaround time
  • Specimen types: nasal wash/swab, nasopharyngeal wash/swab, oropharyngeal swab, BAL, pharynx gargle solution, and saliva
  • Excellent performance with a 100% NPA, PPA
  • Extraction/inhibition control included
The assay simultaneously detects two target sequences in the N gene.
Zertifizierungen: CE-IVD marked.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge