Keep my session open?
Ending In 
The session is expired
Ihre Sitzung ist abgelaufen. Zu Ihrer Sicherheit haben wir Sie abgemeldet.
Möchten Sie sich wieder anmelden?

Due to maintenance activity, Global web site will not be available from 6AM till 1 PM ET on 24th August 2024

J.T.Baker® Endonuclease

Lieferant: AVANTOR PERFORMANCE MATERIALS

BEWERTUNGEN (0)

Gesamtbewertung: 0
Bewertung: von 5

Sortieren nach:


GT20-50EA 11997 EUR InStock
GT20-50 GT20-62 GT20-80
J.T.Baker® Endonuclease
Enzyme
J.T.Baker® Endonuclease Biotech Reagent meets the strictest cGMP standards and is designed for the degradation of both single stranded and double stranded DNA and RNA. J.T.Baker® Endonuclease Biotech Reagent is used to ensure host cell DNA impurities are removed; driving process efficiency by lowering viscosity and preventing aggregation. J.T.Baker® Endonuclease Biotech Reagent is an enzyme based upon the native endonuclease of Serratia marcescens, enabling rapid clearance of residual DNA and RNA during the production and purification of both recombinant proteins and viral vectors. Non-animal origin. Absence of proteolytic activity. The purity of materials in non-negotiable. J.T.Baker® Endonuclease Biotech Reagent acts to degrade and eliminate extraneous genetic material, ensuring the pristine quality of your final product.

  • High purity, bioburden and endotoxin tested
  • Rapid clearance of residual DNA and RNA
  • Easily cleared in downstream processing, detectable by routine analytical methods
  • 0.2 µm filtered solution
  • Various size offerings
  • Proven performance with J.T.Baker® Workflow solutions

Appearance: Passes Test
Identification: Passes Test
Purity by HPLC: ≥ 99%
Activity: ≥ 250 U/µL
Specific Activity: ≥ 1.1 x 10^6 U/mg
Protease: None detected
Total Aerobic Microbial Count: ≤ 10 cfu/ 100,000U
Total Yeast and Mold Count: ≤ 10 cfu/ 100,000U
Endotoxin: ≤ 0.25 EU/1,000U


Order Now

 

PRODUKT

TECHNISCHE DATEN

Bewertung verfassen

BEWERTUNGEN (0)

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge