(±)-Hydroxybupropion-D6 solution, Certified Reference Material (CRM), Ceriliant®, Supelco®
Lieferant: SUPELCO
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H-062-1ML
H-062-1MLEA 259 EUR InStock
H-062-1ML
(±)-Hydroxybupropion-D6 solution, Certified Reference Material (CRM), Ceriliant®, Supelco®
Standards Organische Standards
This stable-labeled internal standard is suitable for quantitation of hydroxybupropion levels in urine, serum, or plasma by LC/MS or GC/MS for urine drug testing, clinical toxicology, forensic analysis, or isotope dilution methods.
(±)-Hydroxybupropion is a major bupropion metabolie in urine. Bupropion is a pharmaceutical used as an antidepressant and a smoking cessation drug sold under the trade name Wellbutrin® and Zyban®.
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7fad341817d1b7b93d9d96b452c5844ac94bd8ad&vid=7fad341817d1b7b93d9d96b452c5844ac94bd8ad&prid=web&clid=SAPDAM
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge