Hartman’s fixative histological tissue fixative, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
BEWERTUNGEN (0)
Gesamtbewertung: 0
Bewertung: von 5
Sortieren nach:
H0290-500ML
H0290-500MLEA 182 EUR InStock
H0290-500ML
Hartman’s fixative histological tissue fixative, Sigma-Aldrich®
Färbemittel Mikroskopie- und Färbereagenzien
Hartman′s fixative, which is also known as Davidson′s Fixative, contains acetic acid, alcohol, and formalin. It is widely used for the preservation of eye, bone marrow, breast, testis, and other tissues for histologic examination
An overnight fixative used for the visualization of lymph nodes in radical dissection specimens. It is helpful in visualizing both both breast and colon specimens by turning lymph nodes white. Moore and Barr used a version of Hartman′s fixative for fixing skin biopsy specimens for study of fine nuclear details. Rat testes for immunohistochemical and morphological comparison were fixed using conventional and modified Hartman′s fixative for 24 hours, while eyes and testes for other analyses were fixed for 48 hours.
Order Now
PRODUKT
TECHNISCHE DATEN
Bewertung verfassen
BEWERTUNGEN (0)
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Oder
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.
Original Product:
Beschreibung Artikelnummer Availability Einheit Ihr Preis Price Per Menge