H25927.18
H25927.09
H25927.18EA
426
EUR
InStock
H25927.18
H25927.09
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate 97%
Ethyl-2-(ethoxymethylene)-4,4,4-trifluoro-3-oxobutyrate
Purity: 97%
CAS No.: 571-55-1
Molecular Formula: C9H11F3O4
Formula Weight: 240.18
Flash Point: 104°(220°F)
Refractive Index: 1.4290
Boiling Point: 80-82°/1mm
MDL No.: MFCD02677683