H25999.03
H25999.09
H25999.03EA
37.1
EUR
InStock
H25999.03
H25999.09
2-Aminoimidazolhemisulfat 98%
2-Aminoimidazolhemisulfat
Purity: 98%
CAS No.: 1450-93-7
Molecular Formula: C6H10N6.0.5H2SO4
Formula Weight: 215.22
Odour: Odourless
Melting Point: ca 270° dec.
Beilstein Reference: 4726750
MDL No.: MFCD00013162