H26002.MD
H26002.03
H26002.MDEA
51.2
EUR
InStock
H26002.MD
H26002.03
2-Amino-5-phenylpyridin 97%
2-Amino-5-phenylpyridine
Purity: 97%
CAS No.: 33421-40-8
Molecular Formula: C11H10N2
Formula Weight: 170.21
Melting Point: 133-137°
MDL No.: MFCD01692452