H26032.MD
H26032.03
H26032.MDEA
75.5
EUR
InStock
H26032.MD
H26032.03
2-Fluorcyclohexanon 96%
2-Fluorocyclohexanone
Purity: 96%
CAS No.: 694-82-6
Molecular Formula: C6H9FO
Formula Weight: 116.13
Storage Temperature: -30°C to -10°C
Flash Point: 71°(160°F)
Refractive Index: 1.4440
Boiling Point: 78°/17mm
MDL No.: MFCD01076476