H26193.03
H26193.06
H26193.03EA
132
EUR
InStock
H26193.03
H26193.06
3-Fluoro-4-nitrobenzyl bromide 97%
3-Fluoro-4-nitrobenzyl bromide
Purity: 97%
CAS No.: 131858-37-2
Molecular Formula: C7H5BrFNO2
Formula Weight: 234.02
Melting Point: 46-48°
UN No.: UN3261
MDL No.: MFCD03094237