H26197.03
H26197.06
H26197.03EA
28.6
EUR
InStock
H26197.03
H26197.06
3-Brom-5-(trifluormethyl)benzaldehyd ≥97%
3-Bromo-5-(trifluoromethyl)benzaldehyde
Purity: 97%
CAS No.: 477535-41-4
Molecular Formula: C8H4BrF3O
Formula Weight: 253.02
Refractive Index: 1.5110