H26212.06
H26212.14
H26212.06EA
78.7
EUR
InStock
H26212.06
H26212.14
4-Brom-2,5-difluorphenol ≥99%
4-Brom-2,5-difluorphenol
Purity: 99%
CAS No.: 486424-36-6
Molecular Formula: C6H3BrF2O
Formula Weight: 208.99
Melting Point: 51-53°
MDL No.: MFCD04973751