H26311.MD
H26311.03
H26311.MDEA
24.8
EUR
InStock
H26311.MD
H26311.03
3-(Trifluormethyl)thiobenzamid ≥97%
3-(Trifluormethyl)thiobenzamid
Purity: 97%
CAS No.: 53515-17-6
Molecular Formula: C8H6F3NS
Formula Weight: 205.20
UN No.: 2811
MDL No.: MFCD01567258