H26627.03
H26627.09
H26627.03EA
55.6
EUR
InStock
H26627.03
H26627.09
4-Fluor-2-methoxybenzonitril ≥97%
4-Fluor-2-methoxybenzonitril
Purity: 97%
CAS No.: 191014-55-8
Molecular Formula: C8H6FNO
Formula Weight: 151.14
Melting Point: 75-77°
UN No.: 3439
MDL No.: MFCD04116335