H26795.03
H26795.06
H26795.03EA
102
EUR
InStock
H26795.03
H26795.06
2-Fluor-5-(trifluormethoxy)benzylbromid ≥97%
2-Fluor-5-(trifluormethoxy)benzylbromid
Purity: 97%
CAS No.: 86256-24-8
Molecular Formula: C8H5BrF4O
Formula Weight: 273.02
Refractive Index: 1.4660