H26815.03
H26815.03EA
137
EUR
InStock
H26815.03
cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylene
cis-Decahydro-2a,4a,6a,8a-tetraazacyclopent[fg]acenaphthylene
CAS No.: 79236-92-3
Molecular Formula: C10H18N4
Formula Weight: 194.28
Melting Point: 29-31°
MDL No.: MFCD09263319