H26879.06
H26879.03
H26879.06EA
109
EUR
InStock
H26879.06
H26879.03
1-Methyl-1H-indazol-3-carbonsäure 97%
1-Methyl-1H-indazole-3-carboxylic acid
Purity: 97%
CAS No.: 50890-83-0
Molecular Formula: C9H8N2O2
Formula Weight: 176.17
Melting Point: ca 218° dec.
MDL No.: MFCD00272569