H26891.03
H26891.03EA
206
EUR
InStock
H26891.03
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II)
Bis(2-methylallyl)-1,5-cyclooctadieneruthenium(II)
CAS No.: 12289-94-0
Molecular Formula: C16H26Ru
Formula Weight: 319.45
Storage Temperature: -30°C to -10°C
Melting Point: 83-86°
UN No.: UN3395
MDL No.: MFCD00216965