H26946.03
H26946.06
H26946.03EA
93.5
EUR
InStock
H26946.03
H26946.06
2,6-Dichlor-4-iodpyridin ≥97%
2,6-Dichloro-4-iodopyridine
Purity: 97%
CAS No.: 98027-84-0
Molecular Formula: C5H2Cl2IN
Formula Weight: 273.89
Melting Point: 161-165°
MDL No.: MFCD07368400