H26957.03
H26957.06
H26957.03EA
64.8
EUR
InStock
H26957.03
H26957.06
(S)-3-(Boc-amino)piperidin 97%
(S)-3-(Boc-amino)piperidin
Purity: 97%
CAS No.: 216854-23-8
Molecular Formula: C10H20N2O2
Formula Weight: 200.28
Melting Point: 122-127°
Optical Rotation: -15.0° (c=1 in MeOH)