H27006.03
H27006.06
H27006.14
H27006.03EA
122
EUR
InStock
H27006.03
H27006.06
H27006.14
(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide
(S,S)-N-(2-Amino-1,2-diphenylethyl)-p-toluenesulphonamide
CAS No.: 167316-27-0
Molecular Formula: C21H22N2O2S
Formula Weight: 366.48
Melting Point: 127-130°
Optical Rotation: +29° (c=1 in chloroform)
MDL No.: MFCD03095684