H27318.14
H27318.06
H27318.14EA
301
EUR
InStock
H27318.14
H27318.06
Methyl-4-oxo-3-piperidincarboxylathydrochlorid 95%
Methyl-4-oxopiperidine-3-carboxylate hydrochloride
Purity: 95%
CAS No.: 71486-53-8
Molecular Formula: C7H11NO3.HCl
Formula Weight: 193.63
Melting Point: ca 175° dec.
Beilstein Reference: 4273241
MDL No.: MFCD00012769