H27444.06
H27444.03
H27444.06EA
86.5
EUR
InStock
H27444.06
H27444.03
3-(Methoxycarbonyl)phenylboronsäure ≥97%
3-(Methoxycarbonyl)phenylboronsäure
Purity: 97%
CAS No.: 99769-19-4
Molecular Formula: C8H9BO4
Formula Weight: 179.97
Melting Point: 136°
MDL No.: MFCD02093046