H27710.14
H27710.03
H27710.14EA
692
EUR
InStock
H27710.14
H27710.03
4-Cyan-3-fluorphenylboronsäure ≥97%
4-Cyan-3-fluorphenylboronsäure
Purity: 97%
CAS No.: 843663-18-3
Molecular Formula: C7H5BFNO2
Formula Weight: 164.93
Melting Point: 300°
MDL No.: MFCD03411549