H27767.06
H27767.03
H27767.06EA
201
EUR
InStock
H27767.06
H27767.03
4-Nitrophenylboronsäure 95%
4-Nitrophenylboronsäure
Purity: 95%
CAS No.: 24067-17-2
Molecular Formula: C6H6BNO4
Formula Weight: 166.93
Melting Point: ca 285° dec.
MDL No.: MFCD00161360