H27847.06
H27847.03
H27847.06EA
61.9
EUR
InStock
H27847.06
H27847.03
3-(3-Fluorphenyl)propionsäure ≥97%
3-(3-Fluorphenyl)propionsäure
Purity: 97%
CAS No.: 458-45-7
Molecular Formula: C9H9FO2
Formula Weight: 168.17
Flash Point: 110°(230°F)
Melting Point: 43-47°
Boiling Point: 125°/10mm
MDL No.: MFCD01310821