H28799.06
H28799.03
H28799.06EA
120
EUR
InStock
H28799.06
H28799.03
(4-Boc-aminophenyl)boronic acid 97%
(4-Boc-aminophenyl)boronic acid
Purity: 97%
CAS No.: 380430-49-9
Molecular Formula: C11H16BNO4
Formula Weight: 237.06
Melting Point: ca 200° dec.
MDL No.: MFCD02093054