H29274.03
H29274.14
H29274.06
H29274.03EA
16.2
EUR
InStock
H29274.03
H29274.14
H29274.06
2,6-Dichlorbenzylamin ≥97%
2,6-Dichlorbenzylamin
Purity: 97+%
CAS No.: 6575-27-5
Molecular Formula: C7H7Cl2N
Formula Weight: 176.05
Odour: Amine-like
UN No.: 2735
MDL No.: MFCD00047928