H29289.06
H29289.03
H29289.06EA
264
EUR
InStock
H29289.06
H29289.03
5-Formyl-2-thiophenboronsäure ≥97%
5-Formyl-2-thiophenboronsäure
Purity: 97%
CAS No.: 4347-33-5
Molecular Formula: C5H5BO3S
Formula Weight: 155.97
Storage Temperature: 2°C to 8°C
Melting Point: ca 124° dec.
MDL No.: MFCD02093666