H30629.03
H30629.06
H30629.03EA
77.4
EUR
InStock
H30629.03
H30629.06
2-Amino-6-fluorpyridin 97%
2-Amino-6-fluorpyridin
Purity: 97%
CAS No.: 1597-32-6
Molecular Formula: C5H5FN2
Formula Weight: 112.11
Odour: Characteristic
Melting Point: 58-62°
UN No.: 2811
MDL No.: MFCD04114200