H31526.03
H31526.06
H31526.09
H31526.03EA
88.4
EUR
InStock
H31526.03
H31526.06
H31526.09
2-Brom-3'-(trifluormethyl)acetophenon ≥98%
2-Brom-3'-(trifluormethyl)acetophenon
Purity: 98%
CAS No.: 2003-10-3
Molecular Formula: C9H6BrF3O
Formula Weight: 267.04
Physical Form: Fused solid
Appearance: Pale yellow
Melting Point: 20-23°
Boiling Point: 69-72°
MDL No.: MFCD03094283