H31974.MD
H31974.03
H31974.MDEA
53.5
EUR
InStock
H31974.MD
H31974.03
2,6-Difluor-4-hydroxybenzaldehyd 95%
2,6-Difluoro-4-hydroxybenzaldehyde
Purity: 95%
CAS No.: 532967-21-8
Molecular Formula: C7H4F2O2
Formula Weight: 158.11
Melting Point: 179-182°
MDL No.: MFCD07772042