H32066.03
H32066.06
H32066.14
H32066.03EA
49.4
EUR
InStock
H32066.03
H32066.06
H32066.14
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol 97%
(1S,2R)-1-Amino-2,3-dihydro-1H-inden-2-ol
Purity: 97%
CAS No.: 126456-43-7
Molecular Formula: C9H11NO
Formula Weight: 149.19
Melting Point: 117-118°
Optical Rotation: -61° (c=0.5 in chloroform)
UN No.: UN3259
MDL No.: MFCD00216655