H32173.03
H32173.06
H32173.03EA
59.4
EUR
InStock
H32173.03
H32173.06
2,2-Difluor-4-pentensäure ≥97%
2,2-Difluoro-4-pentenoic acid
Purity: 97%
CAS No.: 55039-89-9
Molecular Formula: C5H6F2O2
Formula Weight: 136.10
Boiling Point: 56-58°/3mm
UN No.: 3265
MDL No.: MFCD09800642