H32669.06
H32669.14
H32669.22
H32669.06EA
60.6
EUR
InStock
H32669.06
H32669.14
H32669.22
3,5-Dinitrobenzylalkohol ≥98%
3,5-Dinitrobenzylalkohol
Purity: 98%
CAS No.: 71022-43-0
Molecular Formula: C7H6N2O5
Formula Weight: 198.13
Melting Point: 88-91°
Beilstein Reference: 2054388